(4'-Fluoro-[1,1'-biphenyl]-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

51778-500MG 51778-1G 51778-5G 51778-25G
77328-542EA 187.46 USD
77328-542 77352-136 77364-874 77387-024
(4'-Fluoro-[1,1'-biphenyl]-3-yl)methanol ≥97%
(4'-Fluoro-[1,1'-biphenyl]-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₁FO
MW: 202.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05980493
CAS Number: 773871-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR