(3,5-Dichloro-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51777-500MG 51777-1G 51777-5G
77337-416EA 414.06 USD
77337-416 77352-134 77376-414
(3,5-Dichloro-4-methylphenyl)(methyl)sulfane ≥97%
(3,5-Dichloro-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈Cl₂S
Storage Temperature: Ambient
MDL Number: MFCD28787045
CAS Number: 1803837-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR