3-Fluoro-5-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

51770-500MG 51770-1G 51770-5G 51770-25G
77328-540EA 202.91 USD
77328-540 77352-132 77364-872 77387-022
3-Fluoro-5-isopropoxybenzoic acid ≥97%
3-Fluoro-5-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356762
CAS Number: 1369879-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR