4-Fluoro-3-isopropoxy-5-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

51762-500MG 51762-1G 51762-5G
77335-820EA 366.68 USD
77335-820 77352-128 77376-410
4-Fluoro-3-isopropoxy-5-(methylthio)benzaldehyde ≥95%
4-Fluoro-3-isopropoxy-5-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂S
MW: 228.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902971
CAS Number: 2586127-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR