(2,4-Difluoro-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51761-1G 51761-5G 51761-25G
77342-014EA 111.24 USD
77342-014 77364-870 77376-408
(2,4-Difluoro-3-methylphenyl)(methyl)sulfane ≥97%
(2,4-Difluoro-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈F₂S
MW: 174.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28790536
CAS Number: 1807192-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR