1-Bromo-3-chloro-4-ethoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51752-500MG 51752-1G 51752-5G 51752-25G
77328-536EA 166.86 USD
77328-536 77346-206 77364-868 77387-020
1-Bromo-3-chloro-4-ethoxy-2-methylbenzene ≥97%
1-Bromo-3-chloro-4-ethoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564196
CAS Number: 2383566-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR