Methyl 2-chloro-4-ethoxy-3-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

51745-500MG 51745-1G 51745-25G 51745-5G
77328-532EA 306.94 USD
77328-532 77352-122 77387-018 77376-398
Methyl 2-chloro-4-ethoxy-3-fluorobenzoate ≥97%
Methyl 2-chloro-4-ethoxy-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD28503568
CAS Number: 2385083-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR