2-Chloro-4-ethoxy-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

51742-500MG 51742-1G 51742-5G 51742-25G
77328-528EA 219.39 USD
77328-528 77352-118 77364-860 77387-014
2-Chloro-4-ethoxy-3-fluorobenzaldehyde ≥97%
2-Chloro-4-ethoxy-3-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28503572
CAS Number: 1936390-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR