Methyl 2-ethoxy-3-fluoro-4-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
51727-500MG
51727-1G
51727-5G
51727-25G
77328-524EA
281.19
USD
77328-524
77352-102
77364-858
77387-010
Methyl 2-ethoxy-3-fluoro-4-methylbenzoate ≥97%
Methyl 2-ethoxy-3-fluoro-4-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₁H₁₃FO₃ Storage Temperature: Ambient |
MDL Number:
MFCD34564194 CAS Number: 2386736-91-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...