2-Ethoxy-3-fluoro-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

51726-500MG 51726-1G 51726-5G 51726-25G
77328-522EA 225.57 USD
77328-522 77352-100 77364-856 77387-008
2-Ethoxy-3-fluoro-4-methylbenzoic acid ≥97%
2-Ethoxy-3-fluoro-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404000
CAS Number: 2383889-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR