Ethyl 2-(benzyloxy)-3-fluoro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

51720-500MG 51720-1G 51720-5G
77335-402EA 353.29 USD
77335-402 77352-094 77376-382
Ethyl 2-(benzyloxy)-3-fluoro-4-methylbenzoate ≥97%
Ethyl 2-(benzyloxy)-3-fluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD34564193
CAS Number: 2734775-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR