Methyl 2-(benzyloxy)-3-fluoro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

51719-500MG 51719-1G 51719-5G
77328-516EA 331.66 USD
77328-516 77352-092 77376-380
Methyl 2-(benzyloxy)-3-fluoro-4-methylbenzoate ≥97%
Methyl 2-(benzyloxy)-3-fluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₅FO₃
MW: 274.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564192
CAS Number: 2734776-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR