(2-(Benzyloxy)-3-fluoro-4-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

51717-500MG 51717-1G 51717-5G
77328-512EA 334.75 USD
77328-512 77352-088 77376-378
(2-(Benzyloxy)-3-fluoro-4-methylphenyl)methanol ≥97%
(2-(Benzyloxy)-3-fluoro-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₂
MW: 246.277 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756834
CAS Number: 2755723-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR