2-(Benzyloxy)-3-fluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

51716-500MG 51716-1G 51716-25G 51716-5G
77328-510EA 300.76 USD
77328-510 77352-086 77387-000 77376-376
2-(Benzyloxy)-3-fluoro-4-methylbenzaldehyde ≥97%
2-(Benzyloxy)-3-fluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD33403996
CAS Number: 2635937-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR