1-((4-Bromo-2-chlorophenoxy)methyl)-2,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

51715-500MG 51715-1G 51715-5G 51715-25G
77328-508EA 109.18 USD
77328-508 77343-220 77364-848 77376-374
1-((4-Bromo-2-chlorophenoxy)methyl)-2,3-difluorobenzene ≥97%
1-((4-Bromo-2-chlorophenoxy)methyl)-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₈BrClF₂O
MW: 333.556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564191
CAS Number: 2734776-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR