1-((4-Bromo-3-chlorophenoxy)methyl)-2,3-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

51712-500MG 51712-1G 51712-5G 51712-25G
77328-504EA 95.79 USD
77328-504 77342-556 77364-844 77376-370
1-((4-Bromo-3-chlorophenoxy)methyl)-2,3-difluorobenzene ≥95%
1-((4-Bromo-3-chlorophenoxy)methyl)-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₈BrClF₂O
MW: 333.556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564189
CAS Number: 2734778-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR