1-((4-Bromo-2-methylphenoxy)methyl)-2,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

51711-500MG 51711-1G 51711-5G 51711-25G
77328-502EA 140.08 USD
77328-502 77344-718 77364-842 77386-998
1-((4-Bromo-2-methylphenoxy)methyl)-2,3-difluorobenzene ≥97%
1-((4-Bromo-2-methylphenoxy)methyl)-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD19496731
CAS Number: 1284713-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR