(6-Bromo-2-fluoro-3-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

51710-500MG 51710-1G 51710-5G 51710-25G
77328-500EA 187.46 USD
77328-500 77352-084 77364-840 77386-996
(6-Bromo-2-fluoro-3-methoxyphenyl)methanol ≥97%
(6-Bromo-2-fluoro-3-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28141878
CAS Number: 1535997-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR