(4-Fluoro-2-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

51709-1G 51709-5G 51709-25G
77341-332EA 82.4 USD
77341-332 77364-838 77376-368
(4-Fluoro-2-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(4-Fluoro-2-methoxyphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₇FN₂O₂
MW: 252.285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403995
CAS Number: 2624417-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR