(4-Fluoro-2-methoxyphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

51707-1G 51707-5G 51707-25G
77340-710EA 60.77 USD
77340-710 77364-834 77376-366
(4-Fluoro-2-methoxyphenyl)(morpholino)methanone ≥97%
(4-Fluoro-2-methoxyphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄FNO₃
MW: 239.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403993
CAS Number: 2624417-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR