(4-Fluoro-2-methoxyphenyl)(piperidin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

51706-1G 51706-5G 51706-25G
77340-708EA 60.77 USD
77340-708 77364-832 77376-364
(4-Fluoro-2-methoxyphenyl)(piperidin-1-yl)methanone ≥95%
(4-Fluoro-2-methoxyphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆FNO₂
MW: 237.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403992
CAS Number: 2624416-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR