(4-Fluoro-2-methoxyphenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

51705-1G 51705-5G 51705-25G
77340-706EA 60.77 USD
77340-706 77364-830 77376-362
(4-Fluoro-2-methoxyphenyl)(pyrrolidin-1-yl)methanone ≥97%
(4-Fluoro-2-methoxyphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄FNO₂
MW: 223.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403991
CAS Number: 1875060-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR