N-Cyclohexyl-4-fluoro-2-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

51704-1G 51704-5G 51704-25G
77341-110EA 71.07 USD
77341-110 77364-828 77376-360
N-Cyclohexyl-4-fluoro-2-methoxybenzamide ≥97%
N-Cyclohexyl-4-fluoro-2-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD33403990
CAS Number: 2624417-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR