N-Cyclopropyl-4-fluoro-2-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

51703-1G 51703-5G 51703-25G
77341-108EA 71.07 USD
77341-108 77364-826 77376-358
N-Cyclopropyl-4-fluoro-2-methoxybenzamide ≥97%
N-Cyclopropyl-4-fluoro-2-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO₂
MW: 209.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403989
CAS Number: 2624416-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR