4-Fluoro-2-methoxy-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

51702-500MG 51702-1G 51702-5G 51702-25G
77328-496EA 263.68 USD
77328-496 77352-080 77364-824 77386-992
4-Fluoro-2-methoxy-N,N-dimethylbenzamide ≥97%
4-Fluoro-2-methoxy-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO₂
MW: 197.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29091509
CAS Number: 1369777-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR