1,3-Dibromo-5-(tert-butyl)-2-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

51674-1G 51674-5G 51674-25G
77341-230EA 77.25 USD
77341-230 77364-820 77376-354
1,3-Dibromo-5-(tert-butyl)-2-ethoxybenzene ≥97%
1,3-Dibromo-5-(tert-butyl)-2-ethoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₆Br₂O
Storage Temperature: Ambient
MDL Number: MFCD34178605
CAS Number: 2643368-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR