5-Chloro-6-methoxy-2-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

51658-500MG 51658-1G 51658-5G 51658-25G
77328-490EA 184.37 USD
77328-490 77347-212 77364-814 77386-986
5-Chloro-6-methoxy-2-naphthaldehyde ≥97%
5-Chloro-6-methoxy-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₉ClO₂
MW: 220.652 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22192783
CAS Number: 17579-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR