Methyl 5-chloro-6-methoxy-2-naphthoate ≥97%

Supplier: AOBChem USA

Warning

51656-500MG 51656-1G 51656-5G
77335-986EA 372.86 USD
77335-986 77352-074 77376-350
Methyl 5-chloro-6-methoxy-2-naphthoate ≥97%
Methyl 5-chloro-6-methoxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO₃
MW: 250.678 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564188
CAS Number: 2734775-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR