(5-Chloro-6-methoxynaphthalen-2-yl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
51653-500MG
51653-1G
51653-5G
51653-25G
77328-488EA
243.08
USD
77328-488
77352-070
77364-812
77386-984
(5-Chloro-6-methoxynaphthalen-2-yl)(methyl)sulfane ≥95%
(5-Chloro-6-methoxynaphthalen-2-yl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₁₂H₁₁ClOS MW: 238.733 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32902968 CAS Number: 2586126-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...