1,5-Dichloro-3-fluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51652-500MG 51652-1G 51652-5G
77328-486EA 332.69 USD
77328-486 77352-068 77376-346
1,5-Dichloro-3-fluoro-2-methylbenzene ≥97%
1,5-Dichloro-3-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂F
MW: 179.019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236603
CAS Number: 1806351-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR