(3,5-Difluoro-4'-methyl-[1,1'-biphenyl]-4-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51650-500MG 51650-1G 51650-5G
77339-292EA 572.68 USD
77339-292 77352-064 77376-342
(3,5-Difluoro-4'-methyl-[1,1'-biphenyl]-4-yl)(methyl)sulfane ≥97%
(3,5-Difluoro-4'-methyl-[1,1'-biphenyl]-4-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂S
Storage Temperature: Ambient
MDL Number: MFCD32902966
CAS Number: 2586126-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR