3,5-Difluoro-4,4'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

51649-500MG 51649-1G 51649-5G
77328-484EA 346.08 USD
77328-484 77352-062 77376-340
3,5-Difluoro-4,4'-dimethyl-1,1'-biphenyl ≥97%
3,5-Difluoro-4,4'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂
Storage Temperature: Ambient
MDL Number: MFCD33403987
CAS Number: 2624417-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR