(2,6-Dibromo-4-isopropylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

51646-500MG 51646-1G 51646-5G 51646-25G
77328-480EA 136.99 USD
77328-480 77344-596 77364-808 77386-980
(2,6-Dibromo-4-isopropylphenyl)methanol ≥97%
(2,6-Dibromo-4-isopropylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂Br₂O
MW: 308.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564185
CAS Number: 2734777-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR