2-Fluoro-3-methoxy-6-(2,2,2-trifluoroacetamido)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

51628-500MG 51628-1G 51628-5G 51628-25G
77328-474EA 230.72 USD
77328-474 77352-054 77364-804 77386-976
2-Fluoro-3-methoxy-6-(2,2,2-trifluoroacetamido)benzoic acid ≥97%
2-Fluoro-3-methoxy-6-(2,2,2-trifluoroacetamido)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₇F₄NO₄
Storage Temperature: Ambient
MDL Number: MFCD34530137
CAS Number: 2734775-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR