5-Chloro-2-fluoro-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

51616-500MG 51616-1G 51616-5G 51616-25G
77328-470EA 202.91 USD
77328-470 77352-050 77364-800 77386-972
5-Chloro-2-fluoro-3-methoxybenzoic acid ≥97%
5-Chloro-2-fluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₃
MW: 204.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28644342
CAS Number: 1782566-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR