5-Chloro-2-fluoro-1-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51613-500MG 51613-1G 51613-5G
77328-468EA 326.51 USD
77328-468 77352-048 77376-334
5-Chloro-2-fluoro-1-methoxy-3-methylbenzene ≥97%
5-Chloro-2-fluoro-1-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClFO
MW: 174.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751745
CAS Number: 1807167-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR