(5-Chloro-2-fluoro-3-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51611-500MG 51611-1G 51611-5G
77335-682EA 363.59 USD
77335-682 77352-046 77376-332
(5-Chloro-2-fluoro-3-methoxyphenyl)(methyl)sulfane ≥97%
(5-Chloro-2-fluoro-3-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFOS
Storage Temperature: Ambient
MDL Number: MFCD28363342
CAS Number: 1452479-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR