2-Fluoro-3-formyl-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

51589-500MG 51589-1G 51589-5G 51589-25G
77328-460EA 179.22 USD
77328-460 77346-718 77364-796 77386-964
2-Fluoro-3-formyl-5-methoxybenzoic acid ≥97%
2-Fluoro-3-formyl-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₄
Storage Temperature: Ambient
MDL Number: MFCD34756815
CAS Number: 2166812-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR