4-Iodo-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

51572-500MG 51572-1G 51572-5G 51572-25G
77328-454EA 234.84 USD
77328-454 77352-028 77364-790 77386-958
4-Iodo-2-methyl-1,1'-biphenyl ≥97%
4-Iodo-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁I
MW: 294.131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702042
CAS Number: 230961-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR