Methyl(2-methyl-[1,1'-biphenyl]-4-yl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51571-500MG 51571-1G 51571-5G 51571-25G
77328-452EA 150.38 USD
77328-452 77345-352 77364-788 77386-956
Methyl(2-methyl-[1,1'-biphenyl]-4-yl)sulfane ≥97%
Methyl(2-methyl-[1,1'-biphenyl]-4-yl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₄S
MW: 214.326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403975
CAS Number: 2635937-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR