Ethyl 2,4,6-trichloro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

51568-500MG 51568-1G 51568-5G 51568-25G
77328-448EA 145.23 USD
77328-448 77345-128 77364-784 77386-952
Ethyl 2,4,6-trichloro-3-methoxybenzoate ≥97%
Ethyl 2,4,6-trichloro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉Cl₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34564179
CAS Number: 2734776-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR