2-Chloro-3-fluoro-1-methoxy-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

51537-500MG 51537-1G 51537-5G 51537-25G
77328-442EA 217.33 USD
77328-442 77352-022 77364-778 77386-948
2-Chloro-3-fluoro-1-methoxy-4-methylbenzene ≥95%
2-Chloro-3-fluoro-1-methoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD28751724
CAS Number: 1805502-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR