(3-Chloro-2-fluoro-4-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51535-500MG 51535-1G 51535-25G 51535-5G
77328-440EA 304.88 USD
77328-440 77352-020 77386-946 77376-314
(3-Chloro-2-fluoro-4-methoxyphenyl)(methyl)sulfane ≥97%
(3-Chloro-2-fluoro-4-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFOS
Storage Temperature: Ambient
MDL Number: MFCD32902953
CAS Number: 2586126-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR