Ethyl 4-fluoro-2-methoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

51534-500MG 51534-1G 51534-5G
77339-334EA 579.89 USD
77339-334 77352-018 77376-312
Ethyl 4-fluoro-2-methoxy-5-methylbenzoate ≥97%
Ethyl 4-fluoro-2-methoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756810
CAS Number: 2115854-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR