Methyl 4-fluoro-2-methoxy-5-methylbenzoate ≥95%
Supplier: AOBChem USA
|
Warning
|
51533-500MG
51533-1G
51533-5G
77337-412EA
414.06
USD
77337-412
77352-016
77376-310
Methyl 4-fluoro-2-methoxy-5-methylbenzoate ≥95%
Methyl 4-fluoro-2-methoxy-5-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁FO₃ MW: 198.191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31672621 CAS Number: 2121969-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...