(4-Fluoro-2-methoxy-5-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

51531-500MG 51531-1G 51531-5G
77338-094EA 464.53 USD
77338-094 77352-012 77376-306
(4-Fluoro-2-methoxy-5-methylphenyl)methanol ≥97%
(4-Fluoro-2-methoxy-5-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31620764
CAS Number: 1896485-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR