1-Fluoro-4-iodo-5-methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51529-500MG 51529-1G 51529-5G 51529-25G
77328-438EA 194.67 USD
77328-438 77352-010 77364-776 77386-944
1-Fluoro-4-iodo-5-methoxy-2-methylbenzene ≥97%
1-Fluoro-4-iodo-5-methoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD31672622
CAS Number: 2384896-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR