(4-Fluoro-2-methoxy-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51528-500MG 51528-1G 51528-5G
77328-436EA 352.26 USD
77328-436 77352-008 77376-304
(4-Fluoro-2-methoxy-5-methylphenyl)(methyl)sulfane ≥97%
(4-Fluoro-2-methoxy-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
MW: 186.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756809
CAS Number: 2384090-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR