3-Fluoro-4-methyl-2-(methylthio)phenol ≥95%

Supplier: AOBChem USA

Warning

51524-500MG 51524-1G 51524-5G
77336-080EA 375.95 USD
77336-080 77352-006 77376-302
3-Fluoro-4-methyl-2-(methylthio)phenol ≥95%
3-Fluoro-4-methyl-2-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₈H₉FOS
MW: 172.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530128
CAS Number: 2711420-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR