Methyl 2-ethoxy-3,5-bis(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

51517-500MG 51517-1G 51517-5G
77340-366EA 960.99 USD
77340-366 77352-002 77376-298
Methyl 2-ethoxy-3,5-bis(trifluoromethyl)benzoate ≥97%
Methyl 2-ethoxy-3,5-bis(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₀F₆O₃
Storage Temperature: Ambient
MDL Number: MFCD34756804
CAS Number: 2701386-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR